Assay Details
| Gene Symbol: Scn9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290674.1 | NP_001277603.1 | 17 - 18 | 3909 | 72 | |
| NM_001290675.1 | NP_001277604.1 | 17 - 18 | 3876 | 72 | ||
| GenBank mRNA | BC158048.1 | - | 17 - 18 | 3567 | 9088161 | 72 |
| BC172147.1 | - | 17 - 18 | 3600 | 9054817 | 72 | |
More Information
Set Membership: |
|

Scn9a
Genomic Map