Assay Details
| Gene Symbol: Scn9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290675.1 | NP_001277604.1 | 5 - 6 | 1156 | 104 | |
| GenBank mRNA | BC158048.1 | - | 5 - 6 | 847 | 9088161 | 104 |
More Information
Set Membership: |
|

Scn9a
Genomic Map