Assay Details
| Gene Symbol: Anapc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291348.1 | NP_001278277.1 | 1 - 2 | 122 | 94 | |
| NM_027532.4 | NP_081808.2 | 1 - 2 | 122 | 94 | ||
| GenBank mRNA | AK139635.1 | - | 1 - 2 | 114 | 94 | |
| CF166358.1 | - | 1 - 2 | 122 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK139635.1, NM_001291348.1, NM_027532.4 transcripts.
Set Membership: |
|

Anapc15
Genomic Map