Assay Details
| Gene Symbol: Anapc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291348.1 | NP_001278277.1 | 2 - 3 | 171 | 73 | |
| GenBank mRNA | AK139635.1 | - | 2 - 3 | 163 | 73 | |
More Information
Set Membership: |
|

Anapc15
Genomic Map