Assay Details
| Gene Symbol: Anapc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291349.1 | NP_001278278.1 | 1 - 2 | 123 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291349.1 transcripts.
Set Membership: |
|

Anapc15
Genomic Map