Assay Details
| Gene Symbol: Anapc15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291352.1 | NP_001278281.1 | 2 - 3 | 159 | 100 | |
| NM_001291353.1 | NP_001278282.1 | 2 - 3 | 159 | 100 | ||
| NM_027532.4 | NP_081808.2 | 2 - 3 | 201 | 100 | ||
| GenBank mRNA | AK014309.1 | - | 2 - 3 | 118 | 100 | |
| AK018169.1 | - | 1 - 2 | 106 | 100 | ||
| AK135201.1 | - | 2 - 3 | 117 | 100 | ||
| BC006736.1 | - | 2 - 3 | 159 | 3964006 | 100 | |
| BC080730.1 | - | 2 - 3 | 104 | 6507829 | 100 | |
| CF166358.1 | - | 2 - 3 | 201 | 100 | ||
| DV659149.1 | - | 2 - 3 | 171 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014309.1, AK018169.1, AK135201.1, BC006736.1, BC080730.1, NM_001291352.1, NM_001291353.1, NM_027532.4 transcripts.
Set Membership: |
|

Anapc15
Genomic Map