Assay Details
| Gene Symbol: Prss54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016501.1 | - | 2 - 3 | 320 | 93 | |
| BC144859.1 | - | 1 - 2 | 96 | 9053407 | 93 | |
| BC145787.1 | - | 1 - 2 | 96 | 40131090 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC144859.1, BC145787.1 transcripts.
Set Membership: |
|

Prss54
Genomic Map