Assay Details
Gene Symbol: Gm266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033248.3 | NP_001028420.2 | 1 | 521 | 68 | |
GenBank mRNA | AK140917.1 | - | 1 | 484 | 68 | |
AK166394.1 | - | 1 | 495 | 68 | ||
BC145736.1 | - | 1 | 521 | 40131039 | 68 | |
BC145762.1 | - | 1 | 521 | 40131065 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
