Assay Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178874.2 | NP_849205.1 | 3 - 4 | 1773 | 62 | |
| GenBank mRNA | AK157378.1 | - | 3 - 4 | 1886 | 62 | |
| AK158436.1 | - | 2 - 3 | 1135 | 62 | ||
| AK165309.1 | - | 2 - 3 | 1134 | 62 | ||
| AK171635.1 | - | 2 - 3 | 1134 | 62 | ||
| AK172313.1 | - | 2 - 3 | 1134 | 62 | ||
| AK172356.1 | - | 2 - 3 | 1134 | 62 | ||
| AK172541.1 | - | 2 - 3 | 1134 | 62 | ||
| AK172957.1 | - | 3 - 4 | 1628 | 62 | ||
| BC052035.1 | - | 3 - 4 | 1773 | 5695005 | 62 | |
More Information
Set Membership: |
|

Tmcc2
Genomic Map