Assay Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178874.2 | NP_849205.1 | 4 - 5 | 1906 | 102 | |
| GenBank mRNA | AK145948.1 | - | 2 - 3 | 2503 | 102 | |
| AK157378.1 | - | 4 - 5 | 2019 | 102 | ||
| AK158436.1 | - | 3 - 4 | 1268 | 102 | ||
| AK165309.1 | - | 3 - 4 | 1267 | 102 | ||
| AK171635.1 | - | 3 - 4 | 1268 | 103 | ||
| AK172313.1 | - | 3 - 4 | 1267 | 102 | ||
| AK172356.1 | - | 3 - 4 | 1267 | 102 | ||
| AK172382.1 | - | 2 - 3 | 2503 | 102 | ||
| AK172541.1 | - | 3 - 4 | 1267 | 102 | ||
| AK172720.1 | - | 2 - 3 | 2503 | 102 | ||
| AK172957.1 | - | 4 - 5 | 1761 | 102 | ||
| BC052035.1 | - | 4 - 5 | 1906 | 5695005 | 102 | |
More Information
Set Membership: |
|

Tmcc2
Genomic Map