Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 11 - 12 | 1156 | 68 | |
| GenBank mRNA | AK145156.1 | - | 11 - 12 | 1096 | 68 | |
| AK161215.1 | - | 10 - 11 | 1080 | 68 | ||
| AK166337.1 | - | 11 - 12 | 1149 | 68 | ||
| AK172060.1 | - | 11 - 12 | 1154 | 68 | ||
| BC004598.1 | - | 9 - 10 | 877 | 3584958 | 68 | |
More Information
Set Membership: |
|

Sh3bp1
Genomic Map