Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 13 - 14 | 1308 | 61 | |
| GenBank mRNA | AK145156.1 | - | 13 - 14 | 1248 | 61 | |
| AK161215.1 | - | 12 - 13 | 1232 | 61 | ||
| AK166337.1 | - | 13 - 14 | 1301 | 61 | ||
| AK172060.1 | - | 13 - 14 | 1306 | 61 | ||
| BC004598.1 | - | 11 - 12 | 1029 | 3584958 | 61 | |
| X87671.1 | - | 10 - 11 | 1167 | 61 | ||
More Information
Set Membership: |
|

Sh3bp1
Genomic Map