Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 15 - 16 | 1532 | 105 | |
| GenBank mRNA | AK145156.1 | - | 15 - 16 | 1472 | 105 | |
| AK161215.1 | - | 14 - 15 | 1456 | 105 | ||
| AK166337.1 | - | 15 - 16 | 1525 | 105 | ||
| AK172060.1 | - | 15 - 16 | 1530 | 105 | ||
| BC004598.1 | - | 13 - 14 | 1253 | 3584958 | 105 | |
| X87671.1 | - | 12 - 13 | 1391 | 105 | ||
More Information
Set Membership: |
|

Sh3bp1
Genomic Map