Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 1 - 2 | 177 | 102 | |
| GenBank mRNA | AK145156.1 | - | 1 - 2 | 117 | 102 | |
| AK161215.1 | - | 1 - 2 | 178 | 102 | ||
| AK166337.1 | - | 1 - 2 | 170 | 102 | ||
| AK172060.1 | - | 1 - 2 | 174 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_009164.2 transcripts.
Set Membership: |
|

Sh3bp1
Genomic Map