Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 6 - 7 | 588 | 71 | |
| GenBank mRNA | AK145156.1 | - | 6 - 7 | 528 | 71 | |
| AK166337.1 | - | 6 - 7 | 581 | 71 | ||
| AK172060.1 | - | 6 - 7 | 585 | 71 | ||
| BC004598.1 | - | 4 - 5 | 309 | 3584958 | 71 | |
| X87671.1 | - | 3 - 4 | 444 | 71 | ||
More Information
Set Membership: |
|

Sh3bp1
Genomic Map