Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 8 - 9 | 802 | 64 | |
| GenBank mRNA | AK145156.1 | - | 8 - 9 | 742 | 64 | |
| AK161215.1 | - | 7 - 8 | 726 | 64 | ||
| AK166337.1 | - | 8 - 9 | 795 | 64 | ||
| AK172060.1 | - | 8 - 9 | 799 | 64 | ||
More Information
Set Membership: |
|

Sh3bp1
Genomic Map