Assay Details
| Gene Symbol: Sh3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009164.2 | NP_033190.2 | 17 - 18 | 1777 | 79 | |
| GenBank mRNA | AK161215.1 | - | 16 - 17 | 1701 | 79 | |
| AK166337.1 | - | 17 - 18 | 1770 | 79 | ||
More Information
Set Membership: |
|

Sh3bp1
Genomic Map