Assay Details
| Gene Symbol: Hmgxb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178017.1 | NP_821136.1 | 5 - 6 | 1314 | 74 | |
| GenBank mRNA | BC023167.1 | - | 1 - 2 | 56 | 74 | |
| BC048188.1 | - | 6 - 7 | 1429 | 74 | ||
| BC049800.1 | - | 5 - 6 | 1314 | 6509446 | 74 | |
More Information
Set Membership: |
|

Hmgxb4
Genomic Map