Assay Details
| Gene Symbol: Hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC061466.1 | - | 1 - 2 | 319 | 6813540 | 77 |
| BC062905.1 | - | 1 - 2 | 319 | 6813540 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC061466.1, BC062905.1 transcripts.
Set Membership: |
|

Hccs
Genomic Map