Assay Details
| Gene Symbol: Adra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007417.4 | NP_031443.4 | 1 | 1621 | 81 | |
| GenBank mRNA | AK046802.1 | - | 2 | 1415 | 81 | |
| AK140461.1 | - | 2 | 1415 | 81 | ||
| AK141573.1 | - | 1 | 1574 | 81 | ||
| BC138531.1 | - | 1 | 1237 | 8861551 | 81 | |
More Information
Set Membership: |
|

Adra2a
Genomic Map