Assay Details
| Gene Symbol: C1qa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007572.2 | NP_031598.2 | 2 - 3 | 266 | 80 | |
| GenBank mRNA | AK002655.1 | - | 2 - 3 | 192 | 80 | |
| AK136737.1 | - | 2 - 3 | 192 | 80 | ||
| AK150771.1 | - | 2 - 3 | 192 | 80 | ||
| AK159210.1 | - | 2 - 3 | 192 | 80 | ||
| AK159343.1 | - | 2 - 3 | 187 | 80 | ||
| BC002086.1 | - | 2 - 3 | 225 | 3592169 | 80 | |
| BC020096.1 | - | 1 - 2 | 188 | 80 | ||
| BY318890.1 | - | 2 - 3 | 269 | 80 | ||
| X58861.1 | - | 1 - 2 | 189 | 80 | ||
More Information
Set Membership: |
|

C1qa
Genomic Map