Assay Details
| Gene Symbol: Ins2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008387.4 | NP_032413.1 | 1 - 2 | 46 | 81 | |
| GenBank mRNA | AK007612.1 | - | 1 - 2 | 47 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007612.1, NM_008387.4 transcripts.
Set Membership: |
|

Ins2
Genomic Map