Assay Details
| Gene Symbol: Nid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008695.2 | NP_032721.2 | 16 - 17 | 3718 | 90 | |
| GenBank mRNA | AB017202.1 | - | 16 - 17 | 3680 | 90 | |
| AK045306.1 | - | 16 - 17 | 3693 | 90 | ||
| AK051664.1 | - | 16 - 17 | 3698 | 90 | ||
| AK052839.1 | - | 16 - 17 | 3692 | 90 | ||
| AK083629.1 | - | 16 - 17 | 3695 | 90 | ||
| AK128978.1 | - | 13 - 14 | 2840 | 90 | ||
| AK140832.1 | - | 16 - 17 | 3692 | 90 | ||
| AK164258.1 | - | 16 - 17 | 3689 | 90 | ||
| AY074820.1 | - | 16 - 17 | 3679 | 90 | ||
| BC054746.1 | - | 16 - 17 | 3700 | 6405073 | 90 | |
| BC057016.1 | - | 16 - 17 | 3646 | 5719452 | 90 | |
More Information
Set Membership: |
|

Nid2
Genomic Map