Assay Details
| Gene Symbol: Nid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008695.2 | NP_032721.2 | 3 - 4 | 857 | 78 | |
| GenBank mRNA | AK045306.1 | - | 3 - 4 | 831 | 78 | |
| AK052839.1 | - | 3 - 4 | 831 | 78 | ||
| AK083629.1 | - | 3 - 4 | 833 | 78 | ||
| AK140832.1 | - | 3 - 4 | 831 | 78 | ||
| AK164258.1 | - | 3 - 4 | 828 | 78 | ||
| AY074820.1 | - | 3 - 4 | 818 | 78 | ||
| BC050224.1 | - | 3 - 4 | 815 | 78 | ||
| BC054746.1 | - | 3 - 4 | 839 | 6405073 | 78 | |
| BC057016.1 | - | 3 - 4 | 785 | 5719452 | 78 | |
More Information
Set Membership: |
|

Nid2
Genomic Map