Assay Details
| Gene Symbol: Nid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008695.2 | NP_032721.2 | 4 - 5 | 1575 | 107 | |
| GenBank mRNA | AK045306.1 | - | 4 - 5 | 1550 | 107 | |
| AK051664.1 | - | 4 - 5 | 1551 | 107 | ||
| AK052839.1 | - | 4 - 5 | 1549 | 107 | ||
| AK083629.1 | - | 4 - 5 | 1551 | 107 | ||
| AK128978.1 | - | 1 - 2 | 697 | 107 | ||
| AK140832.1 | - | 4 - 5 | 1549 | 107 | ||
| AK164258.1 | - | 4 - 5 | 1546 | 107 | ||
| AY074820.1 | - | 4 - 5 | 1536 | 107 | ||
| BC054746.1 | - | 4 - 5 | 1557 | 6405073 | 107 | |
| BC057016.1 | - | 4 - 5 | 1503 | 5719452 | 107 | |
More Information
Set Membership: |
|

Nid2
Genomic Map