Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 8 - 9 | 1071 | 77 | |
| NM_008860.3 | NP_032886.2 | 11 - 12 | 1271 | 77 | ||
| GenBank mRNA | AB110830.1 | - | 8 - 9 | 1031 | 77 | |
| AK042562.1 | - | 8 - 9 | 1029 | 77 | ||
| AK049959.1 | - | 8 - 9 | 1055 | 77 | ||
| AK147300.1 | - | 8 - 9 | 889 | 77 | ||
| BC057694.1 | - | 3 - 4 | 238 | 77 | ||
| BC072590.1 | - | 8 - 9 | 1071 | 77 | ||
| BC139761.1 | - | 11 - 12 | 1088 | 40109623 | 77 | |
| M94632.1 | - | 11 - 12 | 1088 | 77 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map