Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 11 - 12 | 1407 | 87 | |
| NM_008860.3 | NP_032886.2 | 14 - 15 | 1607 | 87 | ||
| GenBank mRNA | AB110830.1 | - | 11 - 12 | 1367 | 87 | |
| AK042562.1 | - | 11 - 12 | 1365 | 87 | ||
| AK049959.1 | - | 11 - 12 | 1391 | 87 | ||
| AK147300.1 | - | 11 - 12 | 1225 | 87 | ||
| BC057694.1 | - | 6 - 7 | 574 | 87 | ||
| BC072590.1 | - | 11 - 12 | 1407 | 87 | ||
| BC139761.1 | - | 14 - 15 | 1424 | 40109623 | 87 | |
| M94632.1 | - | 14 - 15 | 1424 | 87 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map