Assay Details
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039079.2 | NP_001034168.1 | 14 - 15 | 1696 | 95 | |
| NM_008860.3 | NP_032886.2 | 17 - 18 | 1896 | 95 | ||
| GenBank mRNA | AB110830.1 | - | 14 - 15 | 1656 | 95 | |
| AK042562.1 | - | 14 - 15 | 1654 | 95 | ||
| AK049959.1 | - | 14 - 15 | 1680 | 95 | ||
| AK147300.1 | - | 14 - 15 | 1514 | 95 | ||
| BC057694.1 | - | 9 - 10 | 863 | 95 | ||
| BC072590.1 | - | 14 - 15 | 1696 | 95 | ||
| BC139761.1 | - | 17 - 18 | 1713 | 40109623 | 95 | |
| M94632.1 | - | 17 - 18 | 1713 | 95 | ||
More Information
Set Membership: |
|

Prkcz
Genomic Map