Assay Details
| Gene Symbol: Fbll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004147.3 | NP_001004147.1 | 1 | 922 | 73 | |
| GenBank mRNA | AK140117.1 | - | 1 | 628 | 73 | |
| BC052068.1 | - | 1 | 612 | 5710336 | 73 | |
More Information
Set Membership: |
|

Fbll1
Genomic Map