Assay Details
| Gene Symbol: Tpgs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004361.2 | NP_001004361.2 | 5 - 6 | 764 | 70 | |
| NM_001142697.1 | NP_001136169.1 | 5 - 6 | 510 | 70 | ||
| NM_001142698.1 | NP_001136170.1 | 5 - 6 | 764 | 70 | ||
| GenBank mRNA | AK017728.1 | - | 5 - 6 | 70 | ||
| AK141632.1 | - | 5 - 6 | 581 | 70 | ||
| AK160220.1 | - | 5 - 6 | 560 | 70 | ||
| BC060253.1 | - | 3 - 4 | 316 | 70 | ||
| BC080777.1 | - | 5 - 6 | 520 | 30439612 | 70 | |
| DV655125.1 | - | 5 - 6 | 510 | 70 | ||
More Information
Set Membership: |
|

Tpgs2
Genomic Map