Assay Details
| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB221647.1 | - | 1 - 2 | 47 | 72 | |
| AF090326.1 | - | 131 | 72 | |||
| AK045838.1 | - | 1 - 2 | 162 | 72 | ||
| AK045987.1 | - | 1 - 2 | 162 | 72 | ||
| BC025467.1 | - | 1 - 2 | 854 | 5351959 | 72 | |
| BC031376.1 | - | 2 - 3 | 257 | 5356616 | 72 | |
| CD351822.1 | - | 1 - 2 | 656 | 72 | ||
More Information
Set Membership: |
|

Aebp2
Genomic Map