Assay Details
| Gene Symbol: Shmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009171.2 | NP_033197.2 | 4 - 5 | 724 | 62 | |
| GenBank mRNA | AF237702.1 | - | 4 - 5 | 550 | 62 | |
| AK010439.1 | - | 4 - 5 | 620 | 62 | ||
| AK132342.1 | - | 594 | 62 | |||
| BC026055.1 | - | 589 | 4160579 | 62 | ||
| BQ942592.1 | - | 4 - 5 | 454 | 62 | ||
| X94478.1 | - | 4 - 5 | 550 | 62 | ||
| X94479.1 | - | 4 - 5 | 550 | 62 | ||
More Information
Set Membership: |
|

Shmt1
Genomic Map