Assay Details
| Gene Symbol: Shmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009171.2 | NP_033197.2 | 8 - 9 | 1298 | 101 | |
| GenBank mRNA | AF237702.1 | - | 8 - 9 | 1124 | 101 | |
| AK010439.1 | - | 8 - 9 | 1194 | 101 | ||
| AK132342.1 | - | 1169 | 101 | |||
| BC026055.1 | - | 1163 | 4160579 | 101 | ||
| BQ832130.1 | - | 3 - 4 | 410 | 101 | ||
More Information
Set Membership: |
|

Shmt1
Genomic Map