Assay Details
| Gene Symbol: Sstr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009219.3 | NP_033245.2 | 1 | 1120 | 87 | |
| GenBank mRNA | AK051189.1 | - | 1155 | 87 | ||
| BC138487.1 | - | 1099 | 8861507 | 87 | ||
| BC138488.1 | - | 1099 | 8861508 | 87 | ||
| CB522494.1 | - | 1 | 201 | 87 | ||
More Information
Set Membership: |
|

Sstr4
Genomic Map