Assay Details
| Gene Symbol: Tuba1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009448.4 | NP_033474.1 | 1 - 2 | 531 | 84 | |
| GenBank mRNA | AK145567.1 | - | 1 - 2 | 131 | 84 | |
| AK167654.1 | - | 1 - 2 | 101 | 84 | ||
| AK167759.1 | - | 1 - 2 | 65 | 84 | ||
| BC004745.1 | - | 1 - 2 | 76 | 3583832 | 84 | |
| BC022182.1 | - | 1 - 2 | 141 | 3661220 | 84 | |
| BC026753.1 | - | 1 - 2 | 56 | 5124686 | 84 | |
| BC094022.1 | - | 1 - 2 | 531 | 4162287 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK145567.1, AK167654.1, BC004745.1, BC022182.1, BC026753.1, BC094022.1, NM_009448.4 transcripts.
Set Membership: |
|

Tuba1c
Genomic Map