Assay Details
| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009601.4 | NP_033731.3 | 2 - 3 | 261 | 77 | |
| GenBank mRNA | AK087554.1 | - | 2 - 3 | 240 | 77 | |
| BC023318.1 | - | 2 - 3 | 267 | 4009493 | 77 | |
| CJ176855.1 | - | 2 - 3 | 261 | 77 | ||
| M14537.1 | - | 2 - 3 | 204 | 77 | ||
More Information
Set Membership: |
|

Chrnb1
Genomic Map