Assay Details
| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009601.4 | NP_033731.3 | 3 - 4 | 303 | 70 | |
| GenBank mRNA | AK087554.1 | - | 3 - 4 | 282 | 70 | |
| BC023318.1 | - | 3 - 4 | 309 | 4009493 | 70 | |
| CJ176855.1 | - | 3 - 4 | 303 | 70 | ||
| M14537.1 | - | 3 - 4 | 246 | 70 | ||
More Information
Set Membership: |
|

Chrnb1
Genomic Map