Assay Details
| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009601.4 | NP_033731.3 | 4 - 5 | 409 | 63 | |
| GenBank mRNA | AK087554.1 | - | 4 - 5 | 388 | 63 | |
| BC023318.1 | - | 4 - 5 | 415 | 4009493 | 63 | |
| CJ176855.1 | - | 4 - 5 | 409 | 63 | ||
| M14537.1 | - | 4 - 5 | 352 | 63 | ||
More Information
Set Membership: |
|

Chrnb1
Genomic Map