Assay Details
| Gene Symbol: Chrnb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009601.4 | NP_033731.3 | 5 - 6 | 525 | 101 | |
| GenBank mRNA | AK087554.1 | - | 5 - 6 | 504 | 101 | |
| BC023318.1 | - | 5 - 6 | 531 | 4009493 | 101 | |
| M14537.1 | - | 5 - 6 | 468 | 101 | ||
More Information
Set Membership: |
|

Chrnb1
Genomic Map