Assay Details
| Gene Symbol: Aoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009675.2 | NP_033805.1 | 1 - 2 | 1935 | 59 | |
| GenBank mRNA | AF054831.1 | - | 1 - 2 | 1950 | 59 | |
| AF115411.1 | - | 1 - 2 | 1766 | 59 | ||
| BC080857.1 | - | 1 - 2 | 1774 | 6854465 | 59 | |
More Information
Set Membership: |
|

Aoc3
Genomic Map