Assay Details
| Gene Symbol: Klf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009769.4 | NP_033899.2 | 1 - 2 | 538 | 114 | |
| GenBank mRNA | AB025099.3 | - | 1 - 2 | 512 | 114 | |
| AF079852.1 | - | 1 - 2 | 400 | 114 | ||
| AK051409.1 | - | 1 - 2 | 548 | 114 | ||
| AK087066.1 | - | 1 - 2 | 382 | 114 | ||
| BC006646.1 | - | 1 - 2 | 75 | 3498207 | 114 | |
| BC011307.1 | - | 2 - 3 | 356 | 114 | ||
| BC012958.1 | - | 1 - 2 | 539 | 4208633 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC006646.1 transcripts.
Set Membership: |
|

Klf5
Genomic Map