Assay Details
| Gene Symbol: Klf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009769.4 | NP_033899.2 | 3 - 4 | 1465 | 103 | |
| GenBank mRNA | AB025099.3 | - | 3 - 4 | 1439 | 103 | |
| AF079852.1 | - | 3 - 4 | 1327 | 103 | ||
| AK087066.1 | - | 3 - 4 | 1309 | 103 | ||
| BC011307.1 | - | 4 - 5 | 1285 | 103 | ||
| BC012958.1 | - | 3 - 4 | 1466 | 4208633 | 103 | |
More Information
Set Membership: |
|

Klf5
Genomic Map