Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009797.2 | NP_033927.2 | 9 - 10 | 849 | 129 | |
| GenBank mRNA | AK147049.1 | - | 9 - 10 | 738 | 129 | |
| AK149602.1 | - | 9 - 10 | 735 | 129 | ||
| AK151253.1 | - | 9 - 10 | 746 | 129 | ||
| AK152364.1 | - | 9 - 10 | 734 | 129 | ||
| AK161370.1 | - | 9 - 10 | 851 | 129 | ||
| AK163418.1 | - | 9 - 10 | 735 | 129 | ||
| AK165655.1 | - | 9 - 10 | 736 | 128 | ||
| AK166484.1 | - | 9 - 10 | 733 | 129 | ||
| AK166964.1 | - | 9 - 10 | 738 | 129 | ||
| AK167193.1 | - | 9 - 10 | 736 | 129 | ||
| AK168916.1 | - | 9 - 10 | 737 | 129 | ||
| AK169806.1 | - | 9 - 10 | 735 | 129 | ||
| BC003915.1 | - | 2 - 3 | 141 | 129 | ||
| BC016232.1 | - | 9 - 10 | 738 | 4489604 | 129 | |
| BC085506.1 | - | 9 - 10 | 724 | 6817308 | 129 | |
| U16740.1 | - | 9 - 10 | 718 | 129 | ||
More Information
Set Membership: |
|

Capza1
Genomic Map