Assay Details
| Gene Symbol: Cflar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289704.2 | NP_001276633.1 | 3 - 4 | 631 | 68 | |
| NM_009805.4 | NP_033935.2 | 3 - 4 | 903 | 68 | ||
| NM_207653.5 | NP_997536.1 | 3 - 4 | 903 | 68 | ||
| GenBank mRNA | AK143812.1 | - | 3 - 4 | 896 | 68 | |
| BC029223.1 | - | 3 - 4 | 872 | 4460554 | 68 | |
| BC085105.1 | - | 3 - 4 | 626 | 68 | ||
| U97076.1 | - | 3 - 4 | 569 | 68 | ||
| Y14041.1 | - | 2 - 3 | 481 | 68 | ||
| Y14042.1 | - | 2 - 3 | 481 | 68 | ||
More Information
Set Membership: |
|

Cflar
Genomic Map