Assay Details
| Gene Symbol: Cdk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009870.3 | NP_034000.1 | 7 - 8 | 990 | 72 | |
| GenBank mRNA | AK003804.1 | - | 7 - 8 | 957 | 72 | |
| AK011871.1 | - | 7 - 8 | 978 | 72 | ||
| AK012064.1 | - | 7 - 8 | 959 | 72 | ||
| AK012202.1 | - | 1 - 2 | 76 | 72 | ||
| AK019380.1 | - | 7 - 8 | 943 | 72 | ||
| AK078440.1 | - | 7 - 8 | 944 | 72 | ||
| AK146107.1 | - | 7 - 8 | 957 | 72 | ||
| AK153465.1 | - | 7 - 8 | 990 | 72 | ||
| AK218927.1 | - | 3 - 4 | 277 | 72 | ||
| BC031599.1 | - | 6 - 7 | 1666 | 72 | ||
| BC046336.1 | - | 7 - 8 | 990 | 6312184 | 72 | |
| BC052694.1 | - | 7 - 8 | 985 | 5714971 | 72 | |
| L01640.1 | - | 6 - 7 | 817 | 72 | ||
More Information
Set Membership: |
|

Cdk4
Genomic Map