Assay Details
| Gene Symbol: Dio2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010050.2 | NP_034180.1 | 1 - 2 | 366 | 77 | |
| GenBank mRNA | AF093137.1 | - | 1 - 2 | 224 | 77 | |
| AF096875.1 | - | 1 - 2 | 366 | 77 | ||
| AF177196.1 | - | 1 - 2 | 236 | 77 | ||
| AF177197.1 | - | 1 - 2 | 236 | 77 | ||
| AK032038.1 | - | 1 - 2 | 76 | 77 | ||
| AK045607.1 | - | 1 - 2 | 120 | 77 | ||
| BC125383.1 | - | 1 - 2 | 261 | 40129898 | 77 | |
| BC125385.1 | - | 1 - 2 | 261 | 40129900 | 77 | |
More Information
Set Membership: |
|

Dio2
Genomic Map