Assay Details
| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010058.2 | NP_034188.2 | 4 - 5 | 2014 | 89 | |
| GenBank mRNA | AK086899.1 | - | 3 - 4 | 1576 | 89 | |
| S60312.1 | - | 4 - 5 | 1654 | 89 | ||
More Information
Set Membership: |
|

Dmwd
Genomic Map