Assay Details
| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010213.3 | NP_034343.2 | 2 - 3 | 330 | 59 | |
| GenBank mRNA | AF114382.1 | - | 2 - 3 | 301 | 59 | |
| AF149826.1 | - | 1 - 2 | 128 | 59 | ||
| BC144903.1 | - | 2 - 3 | 294 | 9053451 | 59 | |
| BC145939.1 | - | 2 - 3 | 294 | 40131242 | 59 | |
More Information
Set Membership: |
|

Fhl3
Genomic Map