Assay Details
| Gene Symbol: Khsrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010613.3 | NP_034743.3 | 3 - 4 | 498 | 66 | |
| GenBank mRNA | AK077415.1 | - | 3 - 4 | 466 | 66 | |
| AK156541.1 | - | 3 - 4 | 476 | 66 | ||
| CB247078.1 | - | 2 - 3 | 71 | 66 | ||
More Information
Set Membership: |
|

Khsrp
Genomic Map