Assay Details
| Gene Symbol: Neurod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010895.3 | NP_035025.3 | 1 - 2 | 215 | 63 | |
| GenBank mRNA | AK158767.1 | - | 1 - 2 | 347 | 63 | |
| BC058965.1 | - | 1 - 2 | 164 | 6817440 | 63 | |
| U58471.1 | - | 1 - 2 | 227 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158767.1, BC058965.1, NM_010895.3, U58471.1 transcripts.
Set Membership: |
|

Neurod2
Genomic Map